Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Proto® TorquePlus™ Deep Impact Sockets, ORS Nasco
Supplier: ORS Nasco
577-7328 577-7342 577-7340H 577-07519L 577-7328H 577-7332H 577-7312H 577-7716H 577-7730H 577-07511L 577-7718H 577-7336 577-7336H 577-7714H 577-7326S 577-7711H 577-7330H 577-07514L 577-07517L 577-07524L 577-6506H 577-7709H 577-6520H 577-7316H 577-7332 577-7320 577-7732H 577-6512H 577-07526L 577-6516H 577-7710H 577-7346 577-6508H 577-7334 577-6505MH 577-7346H 577-7312 577-07527L 577-7312S 577-7322S 577-07518L 577-7324H 577-7332S 577-07516L 577-7322H 577-6514H 577-7334H 577-7348 577-07521L 577-07515L 577-7340 577-7720H 577-07513L 577-7324S 577-7728H 577-7314 577-07522L 577-7316S 577-7328S 577-7320S 577-6507H 577-7338H 577-7326 577-7708H 577-07512L 577-07523L 577-7330S 577-7318 577-07510L 577-7314S 577-7336M 577-7344H 577-7322 577-07525L 577-7712H 577-7726H 577-7342H 577-7330 577-7344 577-7324 577-7318H 577-07528L 577-6509H 577-07529L 577-6511H 577-7724H 577-07530L 577-7722H 577-7338 577-07520L 577-7320H 577-7316 577-6510H 577-7314H 577-7318S 577-7326H 577-6518H 577-07509L 577-07532L 577-6911M 577-7348H
300027-784EA 24.27 USD InStock
300027-784 300003-114 300003-112 300027-820 300027-788 300003-110 300027-822 300027-824 300022-846 300028-070 300027-382 300022-966 300027-780 300027-384 300003-118 300027-386 300003-116 300028-066 300028-064 300028-062 300028-060 300027-708 300028-068 300027-796 300027-710 300027-798 300022-972 300027-714 300028-080 300022-978 300003-108 300022-856 300003-106 300027-792 300028-078 300002-678 300028-076 300028-074 300028-072 300002-672 300027-718 300027-762 300027-366 300027-766 300027-800 300022-988 300027-362 300027-804 300027-806 300002-684 300027-808 300027-774 300003-122 300027-776 300003-120 300027-810 300027-812 300022-874 300022-912 300027-370 300027-770 300027-376 300027-814 300027-816 300027-818 300028-058 300027-344 300027-742 300027-348 300022-886 300027-746 300022-802 300031-326 300027-748 300031-318 300027-752 300027-358 300022-930 300022-898 300027-756 300027-758 300022-812 300031-436 300027-352 300031-438 300031-440 300022-780 300027-720 300027-724 300022-786 300031-466 300022-944 300022-826 300031-470 300027-728 300022-794 300027-732 300003-164 300027-736 300022-836 300022-958
Proto® TorquePlus™ Deep Impact Sockets, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Impact Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high-performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in inches (and metric on select models).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty