Would you like to log in again?
Proto® Spline Sockets, #5 Spline, ORS Nasco
Supplier: ORS Nasco
577-09948 577-09936 577-09940 577-09947 577-09916 577-09915 577-09917 577-09931 577-09942 577-09934 577-09924 577-09927 577-09918 577-09932 577-09925 577-09930 577-09941 577-09923 577-09926 577-09919 577-09929 577-09922 577-09920 577-09939 577-09938 577-09933 577-09928 577-09921 577-09935 577-09944
300023-090EA 246.23 USD InStock
300023-090 300023-092 300023-070 300023-094 300023-072 300023-074 300023-096 300023-120 300023-066 300023-088 300023-100 300023-068 300023-102 300023-106 300023-108 300023-080 300023-060 300023-082 300023-062 300023-084 300023-064 300023-086 300023-098 300023-076 300023-110 300023-112 300023-058 300023-114 300023-116 300023-118
Proto® Spline Sockets, #5 Spline, ORS Nasco
Tool Sets
Proto® Spline tools are designed to service high tensile strength fasteners used widely on military and commercial aircraft and in heavy machinery applications where vibration is prevalent
  • Spline sockets help tighten high strength bolts to high torque loads
  • Also used in automotive and industrial areas requiring high performance torque
  • Measurements provided in inches
Spline sockets help lower the force required to turn standard fasteners by allowing the torque load to be moved away from the corners to help prevent fastener rounding.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty