Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Proto® TorquePlus™ Metric Impact Sockets, ORS Nasco
Supplier: ORS Nasco
577-10027M 577-10033M 577-10067M 577-10032M 577-10050M 577-10054M 577-10036M 577-10063M 577-10060M 577-10065M 577-10024M 577-10019M 577-10022M 577-10055M 577-10029M 577-10041M 577-10028M 577-10046M 577-10030M
300023-280EA 79.45 USD InStock
300023-280 300023-172 300023-294 300023-240 300023-186 300023-296 300023-198 300023-220 300023-264 300023-210 300023-288 300023-310 300023-236 300023-302 300023-314 300023-248 300023-226 300023-216 300023-306
Proto® TorquePlus™ Metric Impact Sockets, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Impact Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high-performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in metric scale.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty