Would you like to log in again?
Torqueplus™ Metric 12-Point Offset Box Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
Torqueplus™
577-1064M 577-1061M 577-1079M 577-1077M 577-1051M 577-1053M 577-1065M 577-1073M 577-1057M 577-1068M 577-1069M 577-1072M 577-1058M 577-1050M
300023-350EA 35.05 USD InStock
300023-350 300023-362 300023-340 300023-352 300023-342 300023-364 300023-354 300023-366 300023-344 300023-356 300023-368 300023-346 300023-358 300023-348
Torqueplus™ Metric 12-Point Offset Box Wrenches, Proto®, ORS Nasco
Wrenches
  • TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners
  • Reduces risk of fastener rounding
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Applies more torque to fastener
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty