Would you like to log in again?
Torqueplus™ 12-Point Combination Wrenches - Polish Finish, Proto®, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Torqueplus™
577-1230-T500 577-1214-T500 577-1220-T500 577-1222-T500 577-1218-T500 577-1216-T500 577-1240-T500 577-1212-T500 577-1224-T500 577-1232-T500 577-1236-T500 577-1210-T500 577-1228-T500 577-1234-T500 577-1226-T500
300023-692EA 34.07 USD InStock
300023-692 300023-782 300023-772 300023-596 300023-784 300023-730 300023-776 300023-766 300023-646 300023-712 300023-668 300023-778 300023-746 300023-626 300023-758
Torqueplus™ 12-Point Combination Wrenches - Polish Finish, Proto®, ORS Nasco
Wrenches
TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners.
  • Reduces risk of fastener rounding
  • Applies more torque to fastener
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty