Would you like to log in again?
Torqueplus™ Metric 6-Point Combination Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
Torqueplus™
577-1210MHASD 577-1213MHASD 577-1219MHASD 577-1214MHASD 577-1211MHASD 577-1216MHASD 577-1209MHA 577-1207MHA 577-1217MHASD 577-1208MHA 577-1212MHASD 577-1218MHASD 577-1215MHASD
300023-592EA 28.41 USD InStock
300023-592 300023-572 300023-584 300023-562 300023-642 300023-664 300023-632 300023-676 300023-654 300023-622 300023-688 300023-700 300023-606
Torqueplus™ Metric 6-Point Combination Wrenches, Proto®, ORS Nasco
Wrenches
  • TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners
  • Reduces risk of fastener rounding
  • Applies more torque to fastener
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty