Would you like to log in again?
Torqueplus™ Protoblack™ Metric 12-Point Combination Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Torqueplus™ Protoblack™
577-1224MBASD 577-1207MBA 577-1217MBASD 577-1215MBASD 577-1221MBASD 577-1212MBASD 577-1222MBASD 577-1220MBASD 577-1213MBASD 577-1219MBASD 577-1210MBASD 577-1209MBA 577-1218MBASD 577-1223MBASD 577-1214MBASD 577-1211MBASD 577-1208MBA 577-1216MBASD
300023-708EA 43.88 USD InStock
300023-708 300023-590 300023-570 300023-560 300023-582 300023-640 300023-662 300023-674 300023-652 300023-630 300023-620 300023-686 300023-742 300023-698 300023-736 300023-604 300023-726 300023-718
Torqueplus™ Protoblack™ Metric 12-Point Combination Wrenches, Proto®, ORS Nasco
Wrenches
  • TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners
  • Reduces risk of fastener rounding
  • Applies more torque to fastener
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty