Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Proto® Ergonomics™ Safety Wire Twister Pliers, Straight Jaw, ORS Nasco
Supplier: ORS Nasco
Ergonomics™
577-192 577-195 577-191 577-190
300002-776EA 193.11 USD InStock
300002-776 300002-778 300024-128 300002-774
Proto® Ergonomics™ Safety Wire Twister Pliers, Straight Jaw, ORS Nasco
Pliers
For aviation and other applications where vibration may cause fasteners to back out.
  • Heat-treated cutting edges
  • Holds wire securely and resists kinking
  • Easy access to confined, restricted areas
  • Cuts stainless steel wire
  • Automatic spring return speeds operation and reduces fatigue
Diamond serrated jaws are ideally suited for cutting wire ranging from 0.02 to 0.06" in thickness.
Certifications: FED SPEC. GGG-W-340b; ANSI Spec. B107.18m.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty