Would you like to log in again?
6-Point Heavy-Duty Striking Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-2720SW 577-2756SW 577-2726SW 577-2747SW 577-2738SW 577-2744SW 577-2717SW 577-2741SW 577-2729SW 577-2732SW 577-2750SW 577-2723SW 577-2735SW 577-2762SW
300024-370EA 159.81 USD InStock
300024-370 300024-392 300024-360 300024-380 300024-358 300024-378 300024-376 300024-366 300024-374 300024-386 300024-364 300024-372 300024-384 300024-362
6-Point Heavy-Duty Striking Wrenches, Proto®, ORS Nasco
Wrenches
Ideal for applications where extreme force is needed to loosen or set large fasteners.
  • Large striking surface
  • Efficient transmittal of force to fastener
  • Straight handle design prevents wrench from cocking when struck
  • Box opening remains on fastener when wrench is struck
  • Heavy box walls provide greater torque transmission
  • Short handle permits work in close quarters
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty