Would you like to log in again?
Open End Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-3039 577-3070 577-3026 577-3040 577-3031 577-3035 577-3030 577-3055 577-3021 577-3037 577-3060 577-3080 577-3020 577-3018 577-3034 577-3025 577-3075 577-3016 577-3050 577-3045 577-3051
300024-492EA 46.88 USD InStock
300024-492 300024-470 300024-508 300024-526 300024-504 300024-512 300024-446 300024-458 300024-524 300024-466 300024-444 300024-478 300024-500 300024-442 300024-520 300024-476 300024-454 300024-462 300024-496 300024-472 300024-450
Open End Wrenches, Proto®, ORS Nasco
Wrenches
Increased torque applied to fastener with fewer rounded edges.
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty