Would you like to log in again?
Torqueplus™ 12-Point Double End Flare Nut Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Torqueplus™
577-3772T 577-3764T 577-3768T
300024-846EA 49.72 USD InStock
300024-846 300024-836 300024-840
Torqueplus™ 12-Point Double End Flare Nut Wrenches, Proto®, ORS Nasco
Wrenches
Applies more torque to fastener.
  • Increased torque applied to fastener with fewer rounded edges
  • Improves grip on worn, rounded fasteners
  • Modified square open end provides better fastener grip
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty