Would you like to log in again?
Proto® Ergonomics™ Quick Change Convertible Retaining Ring Pliers, ORS Nasco
Supplier: ORS Nasco
Ergonomics™
577-385L 577-371L 577-376L 577-399L
300024-842EA 49.18 USD InStock
300024-842 300024-876 300024-896 300024-790
Proto® Ergonomics™ Quick Change Convertible Retaining Ring Pliers, ORS Nasco
Pliers
Patented pliers design for quick change from internal to external rings.
  • No screws to tighten
  • Return spring reduces hand fatigue
  • Precision formed and induction hardened fixed tips
With cushioned grips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty