Would you like to log in again?
Proto® 450HS Top Chests, Red, Stanley® Products
Supplier: Stanley
Proto®
577-453427-5RD 577-455027-8RD 577-456627-8RD 577-453427-4RD
300025-160EA 4029.06 USD InStock
300025-160 300025-150 300025-148 300025-162
Proto® 450HS Top Chests, Red, Stanley® Products
Furniture Storage Cabinets
Available in black, blue and yellow, made to order.
  • Made of 14, 16 and 18 gauge steel
  • Standard red powder coat paint finish
  • Heavy duty locks with tubular key
  • Recessed aluminum extruded drawer pulls
  • 100% full extension drawers
Maximum weight capacity for each drawer is 200 lbs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty