Would you like to log in again?
Proto® Super-Duty Drive Pin Punches, ORS Nasco
Supplier: ORS Nasco
577-48116 577-48018 577-48516 577-48038 577-48732 577-48014 577-48332 577-48316 577-48532
300025-614EA 13.23 USD InStock
300025-614 300025-604 300025-610 300025-576 300025-580 300025-618 300025-592 300025-608 300025-582
Proto® Super-Duty Drive Pin Punches, ORS Nasco
Tool Sets
For use in extremely harsh environments.
  • Heat-treated for strength and ductility
  • Modified parabolic striking end for greater resistance to mushrooming and chipping
Certifications: Federal Spec. GGG-P-831e; ANSI Spec. B209.2.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty