Would you like to log in again?
Crowfoot Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
577-4918MCF 577-5344CF 577-5372CF 577-5338CF 577-4962CF 577-4978CF 577-5354CF 577-4958CF 577-4913MCF 577-5364CF 577-4912MCF 577-5348CF 577-4982CF 577-4954CF 577-5360CF 577-5336CF 577-4966CF 577-4980CF 577-4910MCF 577-5378CF 577-5332CF 577-4968CF 577-4919MCF 577-4914CF 577-4928CF 577-4920MCF 577-4917MCF 577-4924CF 577-4916CF 577-5350CF 577-4992CF 577-4964CF 577-4922CF 577-4942CF 577-4990CF 577-4934CF 577-5334CF 577-4924MCF 577-4926CF 577-5356CF 577-5342CF 577-5330CF 577-4930CF 577-4915MCF 577-4920CF 577-4932CF 577-5368CF 577-5352CF 577-4911MCF 577-4972CF 577-5346CF 577-4918CF 577-4946CF 577-4912CF 577-4938CF 577-4922MCF 577-4994CF 577-4986CF 577-5340CF 577-4940CF 577-4948CF 577-4974CF 577-4984CF 577-4950CF 577-4916MCF 577-4970CF 577-4952CF 577-5358CF 577-4988CF 577-4914MCF 577-4944CF 577-5380CF 577-4921MCF 577-4960CF
300025-736EA 89.44 USD InStock
300025-736 300025-738 300025-776 300025-732 300025-656 300025-772 300025-652 300025-730 300026-534 300026-576 300026-494 300026-572 300025-726 300025-848 300025-720 300025-766 300025-722 300025-760 300025-684 300025-680 300025-682 300026-568 300026-526 300026-560 300026-564 300026-484 300025-714 300025-716 300025-676 300025-710 300025-678 300025-712 300025-756 300025-672 300031-068 300025-750 300026-518 300025-674 300025-670 300026-556 300031-072 300026-510 300031-074 300026-592 300025-718 300026-550 300026-594 300031-070 300026-474 300025-748 300025-666 300025-744 300025-700 300025-788 300025-866 300025-668 300025-782 300025-662 300031-058 300025-740 300025-664 300025-862 300025-660 300031-060 300031-062 300026-588 300026-500 300031-064 300031-066 300026-582 300025-706 300026-586 300026-542 300002-896
Crowfoot Wrenches, Proto®, ORS Nasco
Wrenches
  • Anti-Slip Design open end (ASD) has patented locking grooves that help prevent the wrench from slipping off the fastener when turning
  • Crowfoot wrenches are used in hard to reach, restricted access situations where a standard wrench or ratchet cannot reach
  • Full polish finish provides corrosion resistance and easy to clean surfaces
  • Locking hole feature helps keep the crowfoot wrench engaged on the extension during use reducing downtime due to detachment of crowfoot in awkward spaces
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty