Would you like to log in again?
Proto® Super-Duty Roll Pin Punches, ORS Nasco
Supplier: ORS Nasco
577-49018 577-49012 577-49014 577-49316 577-49038 577-49116 577-49332 577-49532 577-49732
300025-636EA 12.94 USD InStock
300025-636 300025-724 300025-638 300025-704 300025-632 300025-698 300025-654 300025-754 300025-634
Proto® Super-Duty Roll Pin Punches, ORS Nasco
Tool Sets
1 piece design.
  • High hardness tip for durability
  • Straighter tip prevents binding
  • Modified parabolic striking end for greater resistance to mushrooming and chipping
Certifications: Federal Spec. GGG-P-831e; ANSI Spec. B209.2.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty