Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Proto® 7-Piece Metric Hex Bit Socket Set, ORS Nasco

Supplier: ORS Nasco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

577-4990-SMA
300025-852EA 144.3 USD InStock
300025-852
Proto® 7-Piece Metric Hex Bit Socket Set, ORS Nasco
Tool Sets
Measurements provided in metric.

  • Made of alloy steel
  • Polished chrome finish
  • Size stamped
  • Short metric hex bits for working in tight spaces

Ordering information: Set includes socket bar and socket clips. Tips sizes are 4 mm, 5 mm, 6 mm, 7 mm, 8 mm, 9 mm, and 10 mm.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty