Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Proto® TorquePlus™ Metric Sockets, ORS Nasco
Supplier: ORS Nasco
577-5426MH 577-5520M 577-5418M 577-5423M 577-5523M 577-5414M 577-5420M 577-5425M 577-5436M 577-5413MH 577-5429MH 577-5530M 577-5420MH 577-5432M 577-5422MH 577-5541M 577-5524M 577-5431MH 577-5430M 577-5425MH 577-5542M 577-5431M 577-5424M 577-5428MH 577-5532M 577-5519M 577-5415M 577-5421MH 577-5421M 577-5438M 577-5529M 577-5521M 577-5419MH 577-5522M 577-5426M 577-5428M 577-5525M 577-5550M 577-5418MH 577-5546M 577-5430MH 577-5413M 577-5429M 577-5427MH 577-5531M 577-5412M 577-5424MH 577-5535M 577-5527M 577-5423MH 577-5411M 577-5536M 577-5417MH 577-5411MH 577-5432MH 577-5417M 577-5528M 577-5410MH 577-5419M 577-5415MH 577-5410M 577-5555M 577-5554M 577-5558M 577-5560M 577-5416M 577-5412MH 577-5416MH 577-5427M 577-5526M 577-5414MH 577-5422M 577-5538M
300027-062EA 150.85 USD InStock
300027-062 300026-816 300027-020 300027-068 300027-024 300026-974 300026-732 300026-776 300026-810 300026-730 300026-774 300026-972 300026-852 300026-978 300026-814 300026-692 300026-970 300026-652 300026-650 300026-694 300027-072 300027-030 300027-034 300026-808 300026-688 300026-766 300026-686 300026-764 300026-962 300026-968 300026-800 300026-760 300027-008 300031-222 300031-224 300027-000 300027-044 300031-228 300027-002 300026-710 300026-798 300026-678 300026-994 300026-676 300026-758 300026-638 300026-636 300026-670 300026-794 300026-750 300026-992 300026-792 300026-672 300027-014 300026-708 300027-052 300031-214 300031-216 300031-218 300026-622 300026-986 300026-984 300026-748 300026-704 300031-220 300026-624 300026-702 300027-060 300026-662 300026-784 300026-660 300026-782 300026-980
Proto® TorquePlus™ Metric Sockets, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in metric.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty