Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Foot Pound Fixed Head Torque Wrenches, Proto®

Supplier: ORS Nasco
577-6023 577-6025
300027-228EA 4232.21 USD InStock
300027-228 300027-230
Foot Pound Fixed Head Torque Wrenches, Proto®
Wrenches
Micrometer torque wrenches with fixed head.

  • Torque wrench is 40" long
  • Weight and overall length are for torque wrench assembled with both extension handles
  • Calibrated to ± 3% of torque reading clockwise, 6% counterclockwise, at 20% to 100% of full scale
  • Certified wrenches to ±2% one direction are available upon request

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty