Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Foot Pound/Meter Kilogram Dial Torque Wrenches, Proto®, ORS Nasco

Supplier: ORS Nasco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

577-6133F 577-6125F 577-6141F 577-6121F 577-6113F
300027-302EA 453.13 USD InStock
300027-302 300027-304 300027-306 300027-308 300027-310
Foot Pound/Meter Kilogram Dial Torque Wrenches, Proto®, ORS Nasco
Wrenches
Dual scale (English/metric) with clockwise and counterclockwise operation - fewer wrenches are needed to complete the job.

  • Accurate to ± 4% of torque reading in both directions at 20% to 100% of full scale
  • Torsion drive design - accuracy not effected by hand position
  • Memory needles are standard on all wrenches except electric light models
  • Electric light and buzzer on 1000 ft.lb. and 2000 ft.lb. models
  • Easy-to-read, high contrast scale

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty