Would you like to log in again?
Inch Pound/Centimeter Kilogram Dial Torque Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-6169F 577-6168F 577-6177F 577-6181F
300027-322EA 471.21 USD InStock
300027-322 300027-312 300027-314 300027-318
Inch Pound/Centimeter Kilogram Dial Torque Wrenches, Proto®, ORS Nasco
Wrenches
Dual scale (English/metric) with clockwise and counterclockwise operation - fewer wrenches are needed to complete the job.
  • Accurate to ± 4% of torque reading in both directions at 20% to 100% of full scale
  • Torsion drive design - accuracy not effected by hand position
  • Memory needles are standard on all wrenches except electric light models
  • Easy-to-read, high contrast scale
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty