Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Torque Multipliers, Proto®, ORS Nasco

Supplier: ORS Nasco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

577-6222 577-6242 577-6252 577-6232 577-6202A 577-6234 577-6212
300027-334EA 5842.86 USD InStock
300027-334 300027-326 300027-338 300091-624 300027-340 300027-330 300027-342
Torque Multipliers, Proto®, ORS Nasco
Wrenches
Use with torque wrenches and heavy-duty power or impact sockets.

  • Standard torque accuracy is +/- 5% of reading
  • Reaction system: tubular reaction bar
  • Gear system: single stage planetary

Accessories information: Proto® torque multipliers provide overload protection with replaceable output square drives that are designed to twist off at 3%-10% over maximum output capacity

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty