Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Proto® TorquePlus™ Impact Sockets, ORS Nasco
Supplier: ORS Nasco
577-7436H 577-7414 577-7412H 577-7432H 577-7442 577-7211H 577-7438 577-7414S 577-7412S 577-6907H 577-7426 577-7420S 577-7418S 577-6920H 577-7428 577-7440H 577-7424H 577-7416 577-7218H 577-7232H 577-7224H 577-6918H 577-7416S 577-7446 577-7418H 577-6914H 577-7432 577-6916H 577-6910H 577-7208H 577-7434S 577-7424 577-7420H 577-7428S 577-7434 577-7422 577-7412 577-7218S 577-7448H 577-7220H 577-7210H 577-7432S 577-7414H 577-6908H 577-7438H 577-7216S 577-7212S 577-7428H 577-7208S 577-7434H 577-7416H 577-7214S 577-7216H 577-7210S 577-6906H 577-7448 577-7444H 577-6911H 577-6912H 577-7430H 577-7444 577-7442H 577-07531 577-7430S 577-7216HF 577-7426S 577-7222H 577-6909H 577-7446H 577-7430 577-7420 577-7422S 577-7228H 577-7436 577-7440 577-7422H 577-7214H 577-07525T 577-7212H 577-7230H 577-7424S 577-7209H 577-7418 577-7426H 577-7208HF 577-7226H 577-07513T
300027-620EA 18.7 USD InStock
300027-620 300027-940 300027-588 300027-424 300027-666 300027-986 300027-866 300027-624 300027-668 300031-368 300027-660 300027-980 300027-584 300027-860 300027-662 300031-450 300031-452 300003-160 300027-902 300027-946 300028-028 300028-026 300028-024 300003-146 300027-630 300027-432 300027-950 300027-554 300003-144 300027-874 300027-996 300027-952 300027-556 300003-142 300027-910 300027-998 300027-956 300027-990 300027-430 300027-992 300027-398 300028-034 300028-032 300028-030 300031-364 300027-638 300027-960 300027-564 300003-136 300027-962 300027-402 300003-134 300027-842 300027-844 300027-966 300022-984 300027-648 300022-822 300027-560 300003-138 300027-882 300027-442 300027-408 300027-606 300028-006 300027-928 300028-004 300028-002 300027-850 300027-454 300027-654 300027-930 300027-458 300027-612 300027-976 300027-856 300003-162 300027-614 300027-416 300022-950 300027-978 300031-454 300027-450 300027-890 300027-572 300027-970 300003-090
Proto® TorquePlus™ Impact Sockets, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Impact Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high-performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in inches.
Certifications: ASME B107.2.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty