Would you like to log in again?
Proto® TorquePlus™ Metric Impact Sockets, ORS Nasco
Supplier: ORS Nasco
577-7215M 577-07534M 577-7418M 577-7421M 577-07546M 577-7425M 577-07541M 577-7412MT 577-7207M 577-07520M 577-7216M 577-7411M 577-7417M 577-69055M 577-7424M 577-7432M 577-7218M 577-07528M 577-7410MT 577-6905M 577-7426M 577-7411MT 577-07529M 577-7412M 577-07527M 577-07531M 577-7420M 577-7416MT 577-07540M 577-6906M 577-6904M 577-07519M 577-07523M 577-7413MT 577-7217M 577-7430MT 577-7213M 577-07550M 577-6907M 577-7415M 577-07533M 577-7427M 577-07526M 577-07521M 577-7206M 577-7419MT 577-6913M 577-7211M 577-7430M 577-7222M 577-7414M 577-7214M 577-7212M 577-07525M 577-7418MT 577-7428M 577-7209M 577-07543M 577-6908M 577-7433M 577-07517M 577-7424MT 577-6915M 577-7434M 577-07542M 577-7423M 577-07530M 577-7422MT 577-7429M 577-6910M 577-7223M 577-7210M 577-7419M 577-07538M 577-6909M 577-7422M 577-07524M 577-7416M 577-6912M 577-7432MT 577-07522M 577-7224M 577-7420MT 577-7220M 577-07535M 577-7409M 577-7410M 577-7219M 577-7408M 577-7221M 577-7415MT 577-7417MT 577-6914M 577-7414MT 577-7413M 577-7436M 577-7208M 577-07532M 577-07536M 577-7421MT
300027-388EA 11.01 USD InStock
300027-388 300022-960 300027-942 300027-426 300027-944 300022-968 300023-004 300023-008 300027-826 300027-948 300027-828 300027-434 300022-974 300027-832 300027-954 300027-558 300027-834 300027-392 300027-394 300027-550 300027-552 300023-014 300027-958 300027-838 300022-860 300027-400 300027-886 300022-980 300022-986 300027-404 300027-646 300027-888 300022-902 300027-880 300027-640 300023-022 300023-026 300023-028 300027-924 300027-926 300027-894 300027-410 300027-652 300022-990 300022-996 300027-898 300027-656 300027-932 300027-934 300022-878 300022-916 300023-040 300027-892 300027-650 300023-032 300023-034 300023-038 300027-418 300027-936 300027-982 300027-586 300027-862 300027-984 300027-622 300027-864 300027-900 300027-988 300027-626 300022-924 300022-890 300027-872 300027-598 300027-994 300027-632 300027-878 300027-912 300027-590 300027-870 300022-936 300027-914 300027-840 300027-566 300027-446 300027-964 300027-448 300027-846 300027-604 300022-946 300027-440 300028-000 300027-968 300027-848 300027-972 300027-610 300027-852 300027-974 300027-854 300022-952 300027-574 300027-616
Proto® TorquePlus™ Metric Impact Sockets, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Impact Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high-performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in metric.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty