Would you like to log in again?
Offset Head Structural Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
577-C907 577-C910 577-C906B 577-C904A 577-C911A 577-C909 577-C903A 577-C911 577-C909A 577-C912 577-C901B 577-C907A 577-C908 577-C905A 577-C905 577-C908A
300028-572EA 138.85 USD InStock
300028-572 300028-584 300028-562 300028-570 300028-582 300028-560 300028-590 300028-580 300028-578 300028-568 300028-576 300028-588 300028-566 300028-574 300028-564 300028-586
Offset Head Structural Wrenches, Proto®, ORS Nasco
Wrenches
Non-reflective ProtoBlack™ finish for use where non-plated products are required.
  • Hardened tapered handle aligns bolt and rivet holes
  • Openings are precision broached for proper fit
  • Offset provides clearance for knuckles and obstructions
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty