Would you like to log in again?
Super Heavy-Duty Offset Slugging Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
577-USN368 577-USN372 577-USN330 577-USN329 577-USN327 577-USN348 577-USN321 577-USN344 577-USN332 577-USN342 577-USN356 577-USN341 577-USN360 577-USN354 577-USN366 577-USN338 577-USN347 577-USN323 577-USN362 577-USN326 577-USN374 577-USN340 577-USN335 577-USN350 577-USN324 577-USN336
300028-728EA 135.7 USD InStock
300028-728 300028-726 300028-748 300028-760 300028-724 300028-746 300028-744 300028-722 300028-766 300028-764 300028-742 300028-720 300028-762 300028-740 300028-718 300028-716 300028-738 300028-736 300028-758 300028-756 300028-734 300028-754 300028-732 300028-730 300028-752 300028-750
Super Heavy-Duty Offset Slugging Wrenches, Proto®, ORS Nasco
Wrenches
  • Heavy box walls provide greater torque transmission
  • Box opening remains on fastener when wrench is struck
  • High strength rectangular design withstands heavy sledging
  • Straight handle resists springing for easier nut turning
  • For close quarter work where hammer or sledge is required to loosen frozen nuts or set large fasteners
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty