Would you like to log in again?
Torqueplus™ Protoblack™ 12-Point Combination Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Torqueplus™ Protoblack™
577-1264B 577-1214BASD 577-1246B 577-1220BASD 577-1228BASD 577-1216BASD 577-1260B 577-1242B 577-1224BASD 577-1240BASD 577-1244B 577-1254B 577-1212BASD 577-1218BASD 577-1248B 577-1258B 577-1234BASD 577-1222BASD 577-1256B 577-1252B 577-1236BASD 577-1230BASD 577-1226BASD
300030-790EA 53.39 USD InStock
300030-790 300030-782 300030-794 300030-780 300030-804 300023-794 300023-798 300023-810 300023-788 300030-786 300030-776 300030-798 300023-812 300030-784 300023-614 300030-774 300023-802 300030-796 300030-802 300030-788 300030-800 300030-778 300023-806
Torqueplus™ Protoblack™ 12-Point Combination Wrenches, Proto®, ORS Nasco
Wrenches
  • TorquePlus™ fastening system drives off fastener body, more easily removing damaged fasteners
  • Reduces risk of fastener rounding
  • Applies more torque to fastener
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove prevents slippage and knuckle injury
  • Improves grip on worn, rounded fasteners
  • Shank is positioned at the center of box end, reduces the tendency of the wrench to cock
  • For access to areas where sockets and attachments will not fit or where greater leverage is required
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty