Would you like to log in again?
Flush Traverse End Cutters, Proto®, ORS Nasco
Supplier: ORS Nasco
577-2834FCMP
300030-964EA 71.95 USD InStock
300030-964
Flush Traverse End Cutters, Proto®, ORS Nasco
Cutters
Flush traverse end cutters are made from hot-drop forged with a high polish finish for strength and cleaning ease.
  • Hardened and tempered jaws for strength and durability
  • For use in limited access or confined ares
Dual leaf spring design helps increase durability while allowing the jaws to be maintained in the open position for quick use. Full flush and semi-flush cutting edges are honed and electronically induction hardened for long life wire cutting.
Certifications: Cushion grip handles on electrical service pliers meet or exceed the electrostatic discharge (ESD) static decay specifications for MIL-PRF-81705D and ESD S.541 (formerly EIA-541).
Confined with ASME B107.25M and B107.38M.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty