Would you like to log in again?
Proto® TorquePlus™ Super Short Sockets, ORS Nasco
Supplier: ORS Nasco
577-5212HS 577-5222HS 577-5224HS 577-5220HS 577-5208HS 577-5216HS 577-5214HS 577-5210HS 577-5206HS 577-5218HS
300031-094EA 11.73 USD InStock
300031-094 300031-096 300031-142 300031-132 300031-100 300031-112 300031-146 300031-124 300031-106 300031-118
Proto® TorquePlus™ Super Short Sockets, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty