Would you like to log in again?
Newton Meter/Inch Pound Dial Torque Wrenches, Proto®, ORS Nasco
Supplier: ORS Nasco
577-6177NMF 577-6121NMF 577-6169NMF
300031-294EA 528.26 USD InStock
300031-294 300027-316 300027-320
Newton Meter/Inch Pound Dial Torque Wrenches, Proto®, ORS Nasco
Wrenches
Dual scale (English/metric) with clockwise and counterclockwise operation - fewer wrenches are needed to complete the job.
  • Accurate to ± 4% of torque reading in both directions at 20% to 100% of full scale
  • Torsion drive design - accuracy not effected by hand position
  • Memory needles are standard on all wrenches except electric light models
  • Easy-to-read, high contrast scale
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty