Would you like to log in again?
Proto® ProtoGrip™ Hi-Strength Magnetic Impact Sockets, ORS Nasco
Supplier: ORS Nasco
577-7211MHF 577-6908MHF 577-6914PF 577-6911MPF 577-7218HF 577-7413MHF 577-6910HF 577-7418PF 577-7215MHF 577-7416PF 577-7208MHF 577-6916HF 577-7213MPF 577-7414PF 577-7416HF 577-6914HF 577-7207MPF 577-7213MHF 577-7217MHF
300027-444EA 24.3 USD InStock
300027-444 300027-412 300027-600 300027-876 300031-442 300031-354 300031-366 300031-322 300031-334 300031-444 300031-378 300031-446 300031-358 300031-448 300027-452 300031-328 300031-380 300031-370 300031-374
Proto® ProtoGrip™ Hi-Strength Magnetic Impact Sockets, ORS Nasco
Tool Sets
6-point magnetic sockets use high strength magnets made from a rare earth magnet alloy/nylon blend
  • ProtoGrip™ Magnetic Sockets require no bushings to hold the magnet in place
  • Allows room for more magnet material, helping increase total magnetic strength
  • Designed to easily pick up heavy fasteners and hold them tightly during use
  • Impact sockets with high impact strength are used in automated and hand impact socket application
  • Sockets are available with either inch or metric measurements
Strong magnetic hold helps increase efficiency when installing fasteners in awkward or hard-to-reach locations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty