Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Proto® TorquePlus™ Thin Wall Deep Impact Sockets, ORS Nasco
Supplier: ORS Nasco
577-7313MHT 577-7320MHT 577-07517-LT 577-7332HT 577-7317MHT 577-07519-LT 577-7314MHT 577-7336HT 577-07524-LT 577-07523-LT 577-7316HT 577-07515-LT 577-7318HT 577-7322HT 577-7310MHT 577-7315MHT 577-7712HT 577-7322MHT 577-7714HT 577-07514-LT 577-07516-LT 577-7334HT 577-7311MHT 577-7312MHT 577-7326HT 577-7330HT 577-07521-LT 577-7328HT 577-7318MHT 577-7722HT 577-7323MHT 577-07518-LT 577-7320HT 577-7321MHT 577-07520-LT 577-7710HT 577-7319MHT 577-07512-LT 577-7324HT 577-7312HT 577-7724HT 577-7720HT 577-7711HT 577-7314HT
300031-428EA 25.25 USD InStock
300031-428 300031-406 300031-408 300031-420 300031-388 300022-866 300022-888 300031-422 300022-922 300022-900 300031-402 300022-828 300031-424 300031-468 300031-404 300022-848 300031-426 300022-804 300031-390 300031-392 300031-394 300031-472 300031-396 300031-430 300031-474 300031-418 300022-876 300022-934 300031-398 300022-858 300031-476 300031-432 300031-410 300031-412 300031-434 300031-478 300031-414 300022-838 300031-416 300031-480 300031-382 300031-482 300031-384 300031-386
Proto® TorquePlus™ Thin Wall Deep Impact Sockets, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Impact Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high-performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in inches (and metric on select models).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty