Would you like to log in again?
Super Junior Vises, Wilton®, ORS Nasco
Supplier: ORS Nasco
Wilton®
825-63247
300034-880EA 237.94 USD InStock
300034-880
Super Junior Vises, Wilton®, ORS Nasco
Vises
These are portable vises, the top jaws are grooved and replaceable.
  • Spindle is fully enclosed
  • Double lockdowns ensure stability
  • Can rotate 45° left or right on base
  • Locking type: threaded
  • Base type: Clamp-On
  • Jaw material: Steel
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty