Would you like to log in again?
Micro-Adjustable 'Click-Type' Torque Wrenches, Wright Tool
Supplier: Wright Tool
875-2477 875-3478
300035-424EA 427.84 USD InStock
300035-424 300035-270
Micro-Adjustable 'Click-Type' Torque Wrenches, Wright Tool
Wrenches
All parts made from allow tool steel for long life accuracy.
  • Positive lock with spring loaded pull down lock ring
  • Push button quick release mechanism (except 6448)
  • Distinct breaking action when the set torque is reached
  • Exposed scale
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty