Would you like to log in again?
Smoke Test™ Brand Smoke Detector Testers, CRC
Supplier: CRC
Smoke Test™
125-02105
300038-724CS 309.46 USD InStock
300038-724
Smoke Test™ Brand Smoke Detector Testers, CRC
Smoke Detectors
Aerosol spray extends user reach up to 6', allows testing to be done while standing safely on the floor.
  • Simulates smoke to provide a true, functional test for smoke detectors
  • Convenient package makes testing easy, increasing the likelihood that detectors will be tested on a regular basis
  • No harmful residue, plastic safe, non-staining
  • Contains no Class I Ozone Depleting Chemicals
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty