Would you like to log in again?
PreSolve® Orange Degreaser, LPS
Supplier: LPS
PreSolve®
428-01405
300039-738EA 695.7 USD InStock
300039-738
PreSolve® Orange Degreaser, LPS
Cleaning Agents Detergents and Surface Cleansers
High performance industrial degreaser which cleans on contact. Safe on all metals and most plastics.
  • Fast acting, deep penetration
  • Natural based
  • Does not contain chlorinated solvents
  • Nonconductive
  • Low residue
  • Inverted spray valve
  • Mild citrus scent
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty