Would you like to log in again?
Blue Moly Compounds, Never-Seez
Supplier: Never-Seez
535-NBBT-16
300040-114EA 72.55 USD InStock
300040-114
Blue Moly Compounds, Never-Seez
Lubricants
Provides excellent protection against extreme pressure and high temperatures
Contains no graphite, lead, or copper, and can be used in applications where these are undesirable or prohibited.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty