Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Absolute™ Digimatic Calipers, Mitutoyo
Supplier: Mitutoyo
Absolute™
504-500-507-10 504-500-506-10
300041-560EA 1237.4 USD InStock
300041-560 300041-038
Absolute™ Digimatic Calipers, Mitutoyo
Calipers
  • Allows direct measurements from power on
  • Zero-setting of the display is also possible at any slider position
  • Special functions which allow it to always remember its measuring position even after the gage has been switched off
Ordering information: It is supplied with the battery with 3.5 Years (20,000 hours).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty