Would you like to log in again?
Weiler® Single Row Heavy-Duty Knot Wire Cup Brush, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-12316 804-13156 804-12406 804-13286P 804-12206 804-12356 804-12856 804-13151 804-13285 804-13257 804-13150 804-13021 804-13286 804-13015 804-12306 804-12276 804-12456 804-12316P 804-13025 804-12866 804-13160 804-13281 804-12376 804-12396 804-12736 804-13163 804-12846 804-13153 804-13258 804-12886 804-12816 804-12476 804-13256 804-13253 804-12376P 804-12256 804-12326 804-12746 804-13284 804-12416 804-13283
300044-128EA 107.45 USD InStock
300044-128 300044-600 300001-007 300044-124 300044-122 300042-652 300044-606 300010-172 300001-005 300042-932 300042-558 300001-006 300044-604 300001-004 300044-602 300042-930 300010-168 300042-562 300042-560 300045-328 300042-648 300049-338 300044-598 300044-574 300044-596 300044-572 300042-642 300010-160 300042-564 300010-161 300010-162 300042-646 300010-163 300010-164 300044-118 300042-644 300042-650 300044-120 300043-344 300043-346 300043-348
Weiler® Single Row Heavy-Duty Knot Wire Cup Brush, ORS Nasco
Brushes
Knot wire cup brush provides heavy-duty, aggressive brushing action, and are best suited for demanding cleaning applications on flat surfaces
These brushes are designed for use with right-angled grinders to remove rust, paint, oxides, and scale.
An ideal tool for removing surface rust and light corrosion from bevel ends prior to welding.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty