Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Air Scribe® Kit, Chicago Pneumatic

Supplier: Chicago Pneumatic

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Air Scribe®
147-93611
300042-971EA 722.38 USD InStock
300042-971
Air Scribe® Kit, Chicago Pneumatic
Pens Stylus and Scribers
Adjustable power for a variety of applications and materials such as metals, plastics, ceramics, and glass.

  • Convertible to a hammer or a light scaler
  • Lightweight for easy operation
  • 5.1 m/s2 vibration
  • 1/4" (NPT) inlet size

Cycle rate is 13,500 blows/min. Air consumption is 1.1 CFM.

Delivery information: Kit includes air filter assembly, air hose, anvil, use with chisels & blanks, carrying case, chisel blank, flat chisel, round nose chisel, stylus, carbide tipped.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty