Would you like to log in again?
Weiler® Crimped Wire Cup Brush, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-14316 804-14206 804-13181 804-13241 804-13244 804-13175 804-13245 804-14606 804-13187 804-13176 804-14126 804-13231 804-14016 804-14166 804-14256 804-14036 804-13245P 804-13188 804-13240 804-14076 804-14616 804-13243 804-14216 804-14026 804-14066
300044-700EA 217.4 USD InStock
300044-700 300042-936 300044-588 300042-934 300044-586 300044-584 300042-938 300010-174 300042-458 300010-175 300010-176 300010-169 300043-354 300044-592 300044-590 300000-990 300044-578 300043-350 300043-352 300044-594 300042-940 300042-566 300000-988 300000-989 300044-580
Weiler® Crimped Wire Cup Brush, ORS Nasco
Brushes
These crimped brushes are designed for use with right-angled grinders to remove rust, paint, oxides, and scale.
  • For light to medium duty, general-purpose applications
  • Very flexible brushing action with consistent performance
  • Features a threaded nut for use on angle grinders
Applications include cleaning, removal, roughening, and deburring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty