Would you like to log in again?
Weiler® Narrow Face Crimped Wire Wheel, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-01805 804-01775 804-00204 804-01705 804-01898 804-01045 804-13081 804-01155 804-01178 804-01675 804-01165 804-01475 804-00154 804-00184 804-00134 804-01145 804-01065 804-00144 804-01258 804-01135 804-00124 804-01075 804-01055 804-01075P
300046-488CS 89.1 USD InStock
300046-488 300044-104 300044-566 300043-086 300044-102 300044-100 300044-110 300044-462 300042-124 300044-108 300010-120 300010-121 300010-122 300010-167 300045-776 300010-124 300010-125 300045-774 300010-126 300045-772 300045-770 300044-098 300044-460 300044-096
Weiler® Narrow Face Crimped Wire Wheel, ORS Nasco
Brushes
These brushes can be used with straight grinders, bench/pedastal grinders, and right-angle grinders.
  • Flexible brushing action
  • Can be mounted in multiples
  • Consistent performance
Brushes are designed for cleaning, light deburring, edge blending, roughening, and finishing applications. Use to remove rust, paint, oxides, and scale.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty