Would you like to log in again?
Weiler® Roughneck® Stringer Bead Wheel, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Weiler®
804-08786 804-09386 804-09856 804-08775 804-08954 804-08776P 804-08926 804-13125 804-08806 804-08796 804-13124 804-13135
300043-868EA 587.42 USD InStock
300043-868 300044-116 300045-322 300043-270 300043-272 300043-782 300043-650 300045-308 300046-492 300043-586 300044-192 300045-306
Weiler® Roughneck® Stringer Bead Wheel, ORS Nasco
Brushes
Designed to remove large burrs and heavy contamination, including rust, paint, oxides, and scale.
  • Very narrow face
  • For hard-to-reach areas, primarily for weld cleaning
  • Suitable for cleaning root and hot passes
Can be used with straight grinders and right-angle grinders.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty