Would you like to log in again?
Weiler® Medium-Face Crimped Wire Wheel, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Weiler®
804-06190 804-06040 804-06060 804-06070 804-06100 804-02325 804-06160 804-02325P 804-06440 804-06090 804-06150 804-06110 804-06170 804-06490 804-02335 804-06180 804-06050 804-06120 804-06080
300041-990EA 243.66 USD InStock
300041-990 300041-992 300044-276 300043-340 300042-554 300042-552 300042-228 300010-130 300010-131 300010-132 300042-556 300043-648 300010-127 300042-550 300042-230 300043-332 300043-334 300043-336 300043-338
Weiler® Medium-Face Crimped Wire Wheel, ORS Nasco
Brushes
Brushes are designed for cleaning, light deburring, edge blending, roughening, and finishing applications.
  • Greater fill density and more aggressive brushing action
  • Can be mounted in multiples
Use to remove rust, paint, oxides, and scale. They can be used with bench/pedastal grinders.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty