Would you like to log in again?
Weiler® Knot Wire Bevel Wheel, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-13406 804-13466 804-13486 804-13436 804-13476
300044-612EA 80.95 USD InStock
300044-612 300044-610 300044-764 300044-608 300010-173
Weiler® Knot Wire Bevel Wheel, ORS Nasco
Brushes
Brushes are designed to remove large burrs and heavy contamination, including rust, paint, oxides, and scale.
  • Maximum aggression for cleaning in corners
  • Heavy-duty brushing action with some flexibility
  • Ideal for cleaning filet welds
Brushes can be used with right-angle grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty