Would you like to log in again?
Weiler® Standard Twist Knot Wire Wheel, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Weiler®
804-08294 804-17687 804-17682 804-08345 804-08264 804-08155 804-13110 804-08095 804-08014 804-08085 804-17689 804-17680 804-08375 804-08004 804-13113P 804-13113 804-13120 804-13106 804-13103 804-08045 804-08135 804-17686 804-08274 804-08189 804-08075 804-08835 804-08106 804-08314 804-08044 804-08325 804-08064 804-08395 804-08105 804-08284 804-17681 804-08034 804-08145 804-08024 804-13106P
300001-009EA 75.65 USD InStock
300001-009 300046-214 300001-008 300046-296 300010-191 300010-192 300010-193 300042-778 300042-776 300042-782 300043-816 300043-818 300045-540 300044-278 300001-010 300042-784 300045-532 300085-292 300010-135 300042-292 300045-534 300010-136 300045-330 300010-137 300010-138 300045-296 300010-139 300045-332 300045-530 300045-298 300085-298 300046-292 300043-862 300046-294 300043-864 300045-536 300043-500 300045-538 300043-866
Weiler® Standard Twist Knot Wire Wheel, ORS Nasco
Brushes
Rugged knot-type construction provides aggressive, high-impact action for severe applications.
  • Use to remove rust, paint, oxides, and scale
  • Can be used with die grinders, straight grinders, bench/pedastal grinders, and right-angle grinders
Designed for cleaning, deburring, and roughening applications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty