Would you like to log in again?
Tiger Disc Angled Style Flap Discs, Weiler
Supplier: Weiler
804-50518 804-50610 804-50562 804-50514 804-50543 804-50519 804-50512 804-50545 804-50502 804-50517 804-50554 804-50644 804-50509 804-50602 804-50516 804-50504 804-50515 804-50605 804-50513 804-50511 804-50505 804-50542 804-50642 804-50520 804-50594 804-50522 804-50544
300044-744EA 33.62 USD InStock
300044-744 300046-312 300046-950 300044-286 300010-270 300010-271 300010-272 300046-756 300010-265 300010-266 300010-267 300010-268 300010-269 300044-472 300047-138 300045-348 300044-138 300046-946 300044-816 300047-070 300010-264 300047-142 300045-930 300047-140 300047-322 300047-586 300045-932
Tiger Disc Angled Style Flap Discs, Weiler
Abrasives
Flap discs with for use on steel, most metals, cast iron or non-ferrous soft metals.
  • Zirconium or aluminum oxide abresive
  • Phenolic or aluminum backing
  • High cut rate and smooth grinding action
  • Phenolic backing prevents disc from scratching the workpiece and offers durability and reinforcement
Ideal for grinding, finishing, weld blending or deburring.
Ordering information: Aluminum oxide abresive, phenolic backing
Zirconium, phenolic backing
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty