Would you like to log in again?
Weiler® Single & Double Spiral Flue Brushes
Supplier: Weiler
804-44125 804-44156 804-44159 804-44036 804-44035 804-44137 804-44133 804-44149 804-44047 804-44135 804-44169 804-44048 804-44037 804-44128 804-44154 804-44131 804-44152 804-44150
300046-520EA 23.95 USD InStock
300046-520 300046-510 300046-522 300046-516 300046-506 300046-518 300046-508 300046-512 300046-502 300046-524 300046-514 300046-504 300045-344 300045-346 300045-340 300045-342 300047-742 300045-338
Weiler® Single & Double Spiral Flue Brushes
Brushes
Designed and engineered from black oil-tempered wire.
  • Cylindrical handle
  • Filled with 0.012" round wire and fitted with 1/4" x 2" NPT pipe nipple
  • Single and double spiral formats available
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty