Would you like to log in again?
Weiler® Banded Crimped Wire End Brushes, ORS Nasco
Supplier: ORS Nasco
804-11113 804-11114 804-11112 804-11103 804-11110
300047-718EA 36.83 USD InStock
300047-718 300048-446 300046-754 300047-716 300047-714
Weiler® Banded Crimped Wire End Brushes, ORS Nasco
Brushes
Crimped brushes are ideal for severe applications to remove rust, paint, oxides, and scale.
  • Banded, solid-end prevents flaring
  • Banded brushes provide shorter trim length for more aggressive brushing action
  • Use for parts that could be damaged by the impact of a knot brush
Use with power drills, drill presses, die grinders, and right-angle air die grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty