Would you like to log in again?
Weiler® Round Power Tube Brushes
Supplier: Weiler
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Weiler®
804-21072 804-21322 804-21078 804-21404 804-21075 804-21080 804-21175 804-21087 804-21203 804-21085 804-21074 804-21143 804-21073 804-21083 804-21149 804-21079 804-21266 804-21282 804-21142 804-21082 804-21076 804-21081 804-21077
300049-346CS 106.92 USD InStock
300049-346 300049-364 300048-950 300049-144 300049-376 300048-962 300048-468 300048-952 300049-142 300010-194 300048-954 300010-195 300049-140 300010-196 300010-197 300048-968 300010-198 300010-199 300050-040 300050-036 300050-048 300050-020 300050-022
Weiler® Round Power Tube Brushes
Brushes
Brushes are ideally suited for deburring, cleaning, and finishing applications.
  • Stainless steel
  • Use fully chucked on smooth or threaded blind holes
  • Single continuous length of stainless steel stem wire
  • Single spiral of crimped wire fill for more flexible brushing action
For use with power drills and flexible shaft tools.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty